missing translation for 'onlineSavingsMsg'
Learn More

CD3 Mouse, PE-Cy7, Clone: SK7, enQuireBio™

Mouse Monoclonal Antibody

Brand:  EnquireBio QAB4-PE7-100Tests

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15931958

  • 451.00 € / 100 tests
Estimated Shipment: 24-01-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

CD3
Monoclonal
5μL/Test
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2
CD3E
Protein A/G Purified
RUO
916
IgG1 κ
Flow Cytometry
SK7
PE-Cyanine7
NP_000724
Mouse
100 Tests
Primary
Store at 2-4°C.
Videos
SDS
Documents
Certificates
Special Offers

Special Offers