missing translation for 'onlineSavingsMsg'
Learn More

CD3 Rat, APC, Clone: 17A2, enQuireBio™

Rat Monoclonal Antibody

Brand:  EnquireBio QAB3-APC-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15931798

  • 191.00 € / 100µg
Estimated Shipment: 28-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

CD3
Monoclonal
0.5mg/mL
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2
Cd3g
Protein A/G Purified
RUO
12502
IgG2b κ
Flow Cytometry
17A2
APC
NP_033980
Rat
100 μg
Primary
Store at 2-4°C.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers