missing translation for 'onlineSavingsMsg'
Learn More

CD3 Rat, FITC, Clone: 17A2, enQuireBio™

Rat Monoclonal Antibody

116.55 € - 156.45 €

Specifications

Antigen CD3
Clone 17A2
Concentration 0.5mg/mL
Applications Flow Cytometry
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15981798
missing translation for 'viewDocuments'
EnquireBio
QAB3-F-25ug
25 μg
116.55 €
25µg
View alternative products
missing translation for 'toSaveUpto' 
»
Please to purchase this item. Need a web account? Register with us today!
15971798
missing translation for 'viewDocuments'
EnquireBio
QAB3-F-100ug
100 μg
156.45 €
100µg
View alternative products
missing translation for 'toSaveUpto' 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Specifications

CD3
0.5mg/mL
Monoclonal
Rat
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2
12502
IgG2b κ
Protein A/G Purified
17A2
Flow Cytometry
FITC
RUO
NP_033980
Cd3g
Primary
Store at 2-4°C.
Videos
SDS
Documents

Documents

Certificates
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.