missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD30 / TNFRSF8 Mouse, Clone: CD30/412, enQuireBio™
Mouse Monoclonal Antibody
Brand: EnquireBio 943-MSM1-P0
Additional Details : Weight : 0.01000kg
Specifications
CD30 / TNFRSF8 | |
Cell surface | |
CD30/412 | |
Unconjugated | |
XP_011540745 | |
Mouse | |
105-120kDa | |
Protein A/G Purified | |
RUO | |
943 | |
IgG1 κ |
Flow Cytometry, Immunofluorescence, Immunohistochemistry | |
Monoclonal | |
200μg/mL | |
10mM PBS with 0.05% BSA and 0.05% azide | |
TNFRSF8 | |
Human CD30 recombinant protein | |
Hodgkin's lymphoma | |
20 μg | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |