missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD30 / TNFRSF8 Mouse, Clone: CD30/412, enQuireBio™
Mouse Monoclonal Antibody
199.00 € - 485.00 €
Specifications
Antigen | CD30 / TNFRSF8 |
---|---|
Clone | CD30/412 |
Concentration | 200μg/mL |
Applications | Flow Cytometry, Immunofluorescence, Immunohistochemistry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15991208
|
EnquireBio
943-MSM1-P1 |
100 μg |
485.00 €
100µg |
Estimated Shipment: 27-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15981208
|
EnquireBio
943-MSM1-P0 |
20 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
CD30 / TNFRSF8 | |
200μg/mL | |
Monoclonal | |
Mouse | |
10mM PBS with 0.05% BSA and 0.05% azide | |
943 | |
Human CD30 recombinant protein | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. | |
105-120kDa |
CD30/412 | |
Flow Cytometry, Immunofluorescence, Immunohistochemistry | |
Unconjugated | |
RUO | |
XP_011540745 | |
TNFRSF8 | |
IgG1 κ | |
Protein A/G Purified | |
Cell surface | |
Hodgkin's lymphoma |