missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CD300e Monoclonal Antibody (UP-H2)
GREENER_CHOICE

Product Code. 17624536
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17624536 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17624536 Supplier Invitrogen™ Supplier No. MA544078

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

This antibody recognizes an extracellular epitope on CD300e / IREM-2, a 32 kDa glycoprotein expressed by mature monocytes and peripheral blood myeloid dendritic cells. Functional application: Stimulation.

CD300e is a member of the glycoprotein family of cell surface proteins expressed on myeloid cells. CD300e interacts with the TYRO protein tyrosine kinase-binding protein and is thought to be involved as an activating receptor. CD300e is expressed on mature myeloid dendritic cells and monocytes, and is down regulated on immature dendritic cells. Further, CD300e interacts with DAP-12, which mediates activating signals. The alloreactive response of naïve T cells has been reported to become enhanced by the activation of CD300e. Diseases associated with CD300e dysfunction include tungiasis and lice infestation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD300e
Applications Flow Cytometry, Functional Assay, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone UP-H2
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene CD300E
Gene Accession No. Q496F6
Gene Alias BC034097; CD300 antigen like family member E; CD300 antigen-like family member E; CD300e; CD300e antigen; CD300e molecule; Cd300le; CLM2; CLM-2; CMRF35A5; CMRF35-A5; CMRF35-like molecule 2; CMRF-35-like molecule-1; Immune receptor expressed on myeloid cells 2; IREM2; IREM-2; PIgR2; PIgR-2; poly-Ig receptor 2; polymeric immunoglobulin receptor 2; TREM5
Gene Symbols CD300E
Host Species Mouse
Immunogen CD300e-HA-transfected cells.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 342510
Target Species Human
Content And Storage 4°C, do not freeze
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.