missing translation for 'onlineSavingsMsg'
Learn More

CD31 Mouse anti-Human, Brilliant Violet 786, Clone: MBC 78.2 (also known as PECAM-1.2; 1.2), BD Optibuild™

Mouse Monoclonal Antibody

Brand:  BD Biosciences 744079

Product Code. 15868759

  • ONLINE EXCLUSIVE PRICE
    404.00 €
    388.12 € / Each
    Save 15.88 €
    3% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

The MBC 78.2 monoclonal antibody recognizes CD31 which is also known as, Platelet endothelial cell adhesion molecule (PECAM-1), platelet GPIIa, or EndoCAM. CD31 is a ñ130 kDa type I transmembrane glycoprotein that belongs to the Ig gene superfamily. CD31 is comprised of an extracellular region with six IgC-like domains, a transmembrane region, and a cytoplasmic domain that contains two immunoreceptor tyrosine-based inhibitory motifs (ITIMs). The MBC 78.2 antibody specifically binds to an epitope located on membrane-proximal, extracellular Ig-like domain 6 of CD31. This epitope remains expressed by activated T cells after enzymatic cleavage and shedding of a soluble extracellular CD31 fragment comprised of Ig-like domains 1 to 5 from cells. In contrast to the MBC 78.2 antibody, the WM59 monoclonal antibody reportedly binds to the extracellular Ig-like domain 2 of CD31. WM59 can thus bind to cells that express intact CD31 but not to cells that express a truncated form CD31 that lacks at least the membrane distal Ig-like domains 1 and 2 of CD31. CD31 has wide tissue distribution and is expressed on platelets, monocytes, granulocytes, some T cell subsets, and at high levels on endothelial cells. This cell adhesion molecule has been implicated in a number of cellular phenomena, including vascular wound healing. angiogenesis, transendothelial migration of leucocytes, and the regulation of T cell responses.

The antibody was conjugated to BD Horizon™ BV786 which is part of the BD Horizon Brilliant™ Violet family of dyes. This dye is a tandem fluorochrome of BD Horizon BV421 with an Ex Max of 405nm and an acceptor dye with an Em Max at 786nm. BD Horizon BV786 can be excited by the violet laser and detected in a filter used to detect Cy™7-like dyes (eg, 780/60nm filter). BV786 is a tandem fluorochrome of BD Horizon BV421 and an acceptor dye with an emission maximum at 786 nm. BV786 offers a bright choice for the 6th detector off the violet laser.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

CD31
MBC 78.2 (also known as PECAM-1.2; 1.2)
Brilliant Violet 786
PECAM-1, PECAM1, EndoCAM, GPIIA'
Human CD31
50 μg
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Monoclonal
0.2mg/mL
Aqueous buffered solution containing ≤0.09% sodium azide.
Mouse
Affinity Purified
RUO
Human
IgG1 κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.