missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CD33 Monoclonal Antibody (WM-53 (WM53)), NovaFluor™ Blue 690, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
185.00 € - 392.00 €
Specifications
| Antigen | CD33 |
|---|---|
| Clone | WM-53 (WM53) |
| Concentration | 0.8 μg/Test |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30055858
|
Invitrogen™
H015T03B09-A |
100 Tests |
392.00 €
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30055193
|
Invitrogen™
H015T02B09-A |
25 Tests |
185.00 €
Pack of 25 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD33 Monoclonal antibody specifically detects CD33 in Human samples. It is validated for Flow CytometrySpecifications
| CD33 | |
| 0.8 μg/Test | |
| Monoclonal | |
| Mouse | |
| Human | |
| P20138 | |
| 945 | |
| Primary | |
| 4°C, store in dark, DO NOT FREEZE! |
| WM-53 (WM53) | |
| Flow Cytometry | |
| NovaFluor Blue 690 | |
| RUO | |
| PBS with 0.09% sodium azide; pH 7.2 | |
| CD33; CD33 antigen; CD33 antigen (gp67); CD33 molecule; CD33 molecule transcript; FLJ00391; gp67; Myeloid cell surface antigen CD33; p67; sialic acid binding Ig-like lectin 3; sialic acid-binding Ig-like lectin 3; SIGLEC3; Siglec-3 | |
| IgG1 κ | |
| Affinity chromatography | |
| CD33 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title