missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD34 Mouse, anti-Human, PE, Clone: 581, STEMCELL Technologies
Mouse Monoclonal Antibody
232.20 € - 513.00 €
Specifications
Antigen | CD34 |
---|---|
Clone | 581 |
Applications | CyTOF, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry |
Classification | Monoclonal |
Conjugate | PE |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17118441
|
Stemcell Technologies
60013PE.1 |
25 Tests |
232.20 €
25 tests |
Estimated Shipment: 16-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17108441
|
Stemcell Technologies
60013PE |
100 Tests |
513.00 €
100 tests |
Estimated Shipment: 16-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- The 581 antibody reacts with human CD34, an ∼105 - 120 kDa type 1 transmembrane glycoprotein expressed on the surface of most human hematopoietic stem and progenitor cells (HSPCs) as well as on mesenchymal stem cells, embryonic fibroblasts, endothelial cells, neurons, and some tumor cell lines
- CD34 is expressed only transiently during hematopoiesis, so the frequency of CD34+ cells is low in bone marrow or cord blood (∼1 - 5%) and very low (∼0.1 - 0.5%) in peripheral blood
- CD34 is a marker used to identify and isolate HSPCs capable of cell engraftment
- This antibody clone has been verified for purity assessments of cells isolated with EasySep™ Human CD34 Positive Selection Kit II (Catalog #17856) and for labeling human mesenchymal cells grown in MesenCult™ Proliferation Kit (Human; Catalog #05411)
Specifications
CD34 | |
CyTOF, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry | |
PE | |
Human | |
Primary |
581 | |
Monoclonal | |
Mouse | |
IgG1 κ |