missing translation for 'onlineSavingsMsg'
Learn More

CD34 Mouse anti-Human, PE-Cy5, Clone: 581, BD

Mouse Monoclonal Antibody

154.08 € - 357.38 €

Specifications

Antigen CD34
Clone 581
Applications Flow Cytometry
Classification Monoclonal
Conjugate PE-Cyanine5
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15841369
View Documents
BD Biosciences
561819
25 Tests
ONLINE EXCLUSIVE PRICE
161.00 €
154.08 € / Each
Save 6.92 €
4% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
15818468
View Documents
BD Biosciences
555823
100 Tests
ONLINE EXCLUSIVE PRICE
372.00 €
357.38 € / Each
Save 14.62 €
3% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The 581 monoclonal antibody specifically binds to CD34, a sialomucin-like type I transmembrane glycoprotein. This single-chain, 105-120 kDa, heavily O-glycosylated protein is expressed on hematopoietic progenitor cells, vascular endothelium, bone marrow stromal cells and embryonic fibroblasts. The cytoplasmic region of the CD34 antigen is a target for phosphorylation by activated protein kinase C suggesting CD34 may play a role in signal transduction. CD34 may also play a role as an adhesion molecule since it binds to CD62E and CD62L. Clone 581 binds to the class III CD34 epitope, it is resistant to neuraminidase, chymopapain and glycoprotease. The 581 antibody blocks reactivity of another anti-CD34 monoclonal antibody, 8G12.

Specifications

Specifications

CD34
Flow Cytometry
PE-Cyanine5
RUO
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
581
Monoclonal
Mouse
Human
IgG1 κ
Affinity Purified
gp105-120; My10; Hematopoietic progenitor cell antigen CD34
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.