missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD35 Antibody (8C12) - Chimeric - Azide and BSA Free, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-52668-0.025MG
This item is not returnable.
View return policy
Description
CD35 Monoclonal specifically detects CD35 in Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Frozen, CyTOF-ready.
Specifications
| CD35 | |
| Monoclonal | |
| Unconjugated | |
| PBS with 0.02% Proclin 300. with 0.02% Proclin 300 | |
| CR1 | |
| Purified murine CR1 protein. | |
| 0.025 mg | |
| Primary | |
| Recognises an epitope on CD35 (CR1, 190 kDa), but not on CD21 (CR2, 145-150 kDa), which are alternative splice variants of primary Cr2 gene transcripts. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Flow Cytometry, Immunoprecipitation, Immunohistochemistry (Frozen), CyTOF | |
| 8C12 | |
| Western Blot 1:100 - 1:2000, Flow Cytometry 1:10 - 1:1000, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Frozen 1:10 - 1:500, CyTOF-ready | |
| complement component (3b/4b) receptor 1 (Knops blood group), KN | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| 1378 | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction