Learn More
CD35 Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 30057-1-AP-20UL
 View more versions of this product
Product Code. 18054505
  • 145.00 € / 20µL
Description
The CD35 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human CD35. This antibody recognizes human antigen. The CD35 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Specifications
CD35
600 μg/mL
Rabbit
Primary
Human
150 kDa
IgG
Western Blot, ELISA
C3b/C4b receptor, C3BR, CD35, Complement receptor type 1, CR1, KN
20 μL
1378
Store at -20°C. Stable for one year after shipment.