Learn More
CD36 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 82 publications
Brand:  Novus Biologicals NB400-144SS
 View more versions of this product
Product Code. 18719873
  • 314.00 € / 25µL
Description
CD36 Polyclonal specifically detects CD36 in Human, Mouse, Rat, Porcine, Bovine, Primate, Rabbit samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CD36/SR-B3
Polyclonal
Unconjugated
PBS with 0.02% Sodium Azide
CD36 antigen, CD36 molecule (thrombospondin receptor), FA6-152, FATCHDS7, Fatty acid translocase, Glycoprotein IIIb, GP3Bthrombospondin receptor), GPIIIB, Leukocyte differentiation antigen CD36, PAS IV, PAS-4, Platelet collagen receptor, platelet glycoprotein 4, Platelet glycoprotein IV, scavenger receptor class B, member 3, Thrombospondin receptor
Rabbit
75 kDa
0.025 mL
Cancer, Cellular Markers, Endothelial Cell Markers, Hematopoietic Stem Cell Markers, Immunology, Lipid and Metabolism, Stem Cell Markers
948
Human, Mouse, Rat, Pig, Bovine, Primate, Rabbit
IgG
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
1 mg/mL
Western Blot 1:500-1:2000, Immunohistochemistry 1:200-1:400, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunohistochemistry-Paraffin 1:200-1:400, Immunohistochemistry-Frozen reported in scientific literature (PMID 24531551)
P16671
CD36
A synthetic peptide mapping to a region of human CD36 between residues 100-200. [UniProt# P16671]
Affinity Purified
RUO
Primary
Human, bovine, mouse and monkey. Porcine reactivity reported in scientific literature (PMID: 23727393). Immunogen sequence has 95% identity to rat.
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.