Learn More
CD36 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 82 publications
Brand:  Novus Biologicals NB400-144
 View more versions of this product
Product Code. 18797473
  • 637.00 € / 0.10mL
Description
CD36 Polyclonal specifically detects CD36 in Human, Mouse, Rat, Porcine, Bovine, Primate, Rabbit samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Polyclonal
CD36
75 kDa
RUO
IgG
Western Blot 1:500-1:2000, Immunohistochemistry 1:200-1:400, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunohistochemistry-Paraffin 1:200-1:400, Immunohistochemistry-Frozen reported in scientific literature (PMID 24531551)
A synthetic peptide mapping to a region of human CD36 between residues 100-200. [UniProt# P16671]
Affinity Purified
Pig