Learn More
Novus Biologicals™ CD37 Antibody (AGS67E) - IgG2SA, Novus Biologicals™
Human Monoclonal Antibody
Brand:  Novus Biologicals™ NBP3-28746-100ug
 View more versions of this product
Product Code. 30047456
  • 349.00 € / 100µg
Description
CD37 Monoclonal antibody specifically detects CD37 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CD37
Monoclonal
Unconjugated
P11049
Human
Protein A purified
RUO
Primary
Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex.
Store at -20°C in powder form. Store at -80°C once reconstituted.
IgG2
Flow Cytometry, ELISA, Functional Assay
AGS67E
Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2)
CD37 antigentetraspanin-26, CD37 molecule, cell differentiation antigen 37, GP52-40, leukocyte antigen CD37, MGC120234, Tetraspanin-26, Tspan-26, TSPAN26leukocyte surface antigen CD37
TSPAN26/ CD37
100 μg
Cellular Markers, Immunology
951
Human
Purified