Learn More
CD37 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-33969
This item is not returnable.
View return policy
Description
CD37 Polyclonal specifically detects CD37 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Polyclonal | |
| RUO | |
| IgG |
| This antibody was developed against a recombinant protein corresponding to amino acids: PCSCYNLSATNDSTILDKVILPQLSRLGHLARSRHSADICAVPAESHIYREGCAQGLQKWLHNN | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |