missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CD37 Recombinant Human Monoclonal Antibody (9H6)
GREENER_CHOICE

Product Code. 30535078
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30535078 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30535078 Supplier Invitrogen™ Supplier No. MA560781

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Human Recombinant Monoclonal Antibody

CD37 Recombinant Monoclonal Antibody for IHC (P), ELISA

The protein encoded by this gene is a member of the transmembrane 4 superfamily, also known as the tetraspanin family. Most of these members are cell-surface proteins that are characterized by the presence of four hydrophobic domains. The proteins mediate signal transduction events that play a role in the regulation of cell development, activation, growth and motility. This encoded protein is a cell surface glycoprotein that is known to complex with integrins and other transmembrane 4 superfamily proteins. It may play a role in T-cell-B-cell interactions. Alternate splicing results in multiple transcript variants encoding different isoforms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD37
Applications ELISA, Immunohistochemistry (Paraffin)
Classification Recombinant Monoclonal
Clone 9H6
Concentration 3 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene CD37
Gene Accession No. P11049
Gene Alias CD37; CD37 antigen; CD37 molecule; cell differentiation antigen 37; GP52-40; Leukocyte antigen CD37; leukocyte surface antigen CD37; tetraspanin-26; Tspan26; Tspan-26
Gene Symbols CD37
Host Species Human
Immunogen Recombinant Human CD37 protein.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 951
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.