missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD38 Mouse, APC, Clone: HIT2, enQuireBio™
Mouse Monoclonal Antibody
131.00 € - 312.00 €
Specifications
Antigen | CD38 |
---|---|
Clone | HIT2 |
Concentration | 5μL/Test |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15991908
|
EnquireBio
QAB38-APC-25TESTS |
25 Tests |
131.00 €
25 tests |
Estimated Shipment: 13-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15981908
|
EnquireBio
QAB38-APC-100TESTS |
100 Tests |
312.00 €
100 tests |
Estimated Shipment: 13-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
CD38 | |
5μL/Test | |
Monoclonal | |
Mouse | |
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2 | |
952 | |
IgG1 κ | |
Protein A/G Purified |
HIT2 | |
Flow Cytometry | |
APC | |
RUO | |
NP_001766 | |
CD38 | |
Primary | |
Store at 2-4°C. |