missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CD39 Monoclonal Antibody (eBioA1 (A1)), NovaFluor™ Yellow 755, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
178.00 € - 392.00 €
Specifications
| Antigen | CD39 |
|---|---|
| Clone | eBioA1 (A1) |
| Concentration | 0.6 μg/Test |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30055793
|
Invitrogen™
H032T03Y08-A |
100 Tests |
392.00 €
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30054895
|
Invitrogen™
H032T02Y08-A |
25 Tests |
178.00 €
Pack of 25 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD39 Monoclonal antibody specifically detects CD39 in Human samples. It is validated for Flow CytometrySpecifications
| CD39 | |
| 0.6 μg/Test | |
| Monoclonal | |
| Mouse | |
| Human | |
| P49961 | |
| 953 | |
| Primary | |
| 4°C, store in dark, DO NOT FREEZE! |
| eBioA1 (A1) | |
| Flow Cytometry | |
| NovaFluor Yellow 755 | |
| RUO | |
| PBS with 0.09% sodium azide; pH 7.2 | |
| 2610206B08Rik; AA408691; ATPDase; Cd39; CD39 antigen; DKFZp686D194; DKFZp686I093; ecto-apyrase; Ecto-ATP diphosphohydrolase 1; ecto-ATPase 1; ecto-ATPDase 1; Ectonucleoside triphosphate diphosphohydrolase 1; ENTPD1; FLJ40921; FLJ40959; Lymphoid cell activation antigen; NTPDase 1; NTPDase-1; SPG64 | |
| IgG1 κ | |
| Affinity chromatography | |
| ENTPD1 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title