missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD3e Armenian Hamster, Clone: 145-2C11, enQuireBio™
Armenian Hamster Monoclonal Antibody
131.25 € - 327.60 €
Specifications
| Antigen | CD3e |
|---|---|
| Clone | 145-2C11 |
| Concentration | 0.5mg/mL |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15931628
|
EnquireBio
QAB2-100ug |
100 μg |
131.25 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15941628
|
EnquireBio
QAB2-1mg |
1 mg |
327.60 €
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
| CD3e | |
| 0.5mg/mL | |
| Monoclonal | |
| Armenian Hamster | |
| 10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, pH 7.2 | |
| 12501 | |
| IgG | |
| Protein A/G Purified |
| 145-2C11 | |
| Flow Cytometry | |
| Unconjugated | |
| RUO | |
| XP_006510029 | |
| Cd3e | |
| Primary | |
| Store at 2-4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title