missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD4 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
184.00 € - 433.00 €
Specifications
| Antigen | CD4 |
|---|---|
| Concentration | 1.0 mg/ml |
| Dilution | Western Blot 1 - 2 ug/ml, Flow Cytometry 2 - 3 ug/ml, Immunohistochemistry 1:200, Immunocytochemistry/Immunofluorescence 1 - 2 ug/ml, Immunohistochemistry-Paraffin 1:200, Flow (Intracellular) 2 - 3 ug/ml |
| Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Flow Cytometry |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18304662
|
Bio-Techne
NBP3-18057SS |
0.025 mg |
184.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18013494
|
Novus Biologicals
NBP3-18057 |
0.1 mg |
433.00 €
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD4 Polyclonal antibody specifically detects CD4 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Flow (Intracellular)Specifications
| CD4 | |
| Western Blot 1 - 2 ug/ml, Flow Cytometry 2 - 3 ug/ml, Immunohistochemistry 1:200, Immunocytochemistry/Immunofluorescence 1 - 2 ug/ml, Immunohistochemistry-Paraffin 1:200, Flow (Intracellular) 2 - 3 ug/ml | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| biosimilar, biosimilars, CD_antigen: CD4, CD4, CD4 antigen, CD4 antigen (p55), CD4 molecule, CD4 receptor, CD4mut, T-cell surface antigen T4/Leu-3, T-cell surface glycoprotein CD4 | |
| Partial recombinant protein made to an N-terminal portion of human CD4 (amino acids 26-390) [UniProt P01730] | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| 1.0 mg/ml | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Flow Cytometry | |
| Unconjugated | |
| Rabbit | |
| Adaptive Immunity, Cell Biology, Cellular Markers, Cytokine Research, Immunology, Innate Immunity, Regulatory Immunology, Signal Transduction, Stem Cell Markers | |
| PBS | |
| 920 | |
| IgG | |
| Immunogen affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title