missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD4 Mouse anti-Human, BUV563, Clone: OKT4, BD Biosciences™
Description
CD4 Monoclonal antibody specifically detects CD4 in Human samples. It is validated for Flow Cytometry
Specifications
Specifications
| Antigen | CD4 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | OKT4 |
| Conjugate | Brilliant Ultraviolet 563 |
| Formulation | Aqueous buffered solution containing ≤0.09% sodium azide. |
| Gene Alias | CD4 antigen (p55); T-cell surface antigen T4/Leu-3 |
| Host Species | Mouse |
| Immunogen | Sheep Erythrocyte Rosette-purified Human T Cells |
| Quantity | 50 μg |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?