missing translation for 'onlineSavingsMsg'
Learn More

CD4 Mouse, FITC, Clone: RPA-T4, enQuireBio™

Mouse Monoclonal Antibody

116.55 € - 565.95 €

Specifications

Antigen CD4
Clone RPA-T4
Concentration 5μL/Test
Applications Flow Cytometry
Classification Monoclonal
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15911518
View Documents
EnquireBio
QAB12-F-25Tests
25 Tests
116.55 €
25 tests
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
15901518
View Documents
EnquireBio
QAB12-F-100Tests
100 Tests
156.45 €
100 tests
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
15921518
View Documents
EnquireBio
QAB12-F-500Tests
500 Tests
565.95 €
500 tests
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Specifications

CD4
5μL/Test
Monoclonal
Mouse
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2
920
IgG1 κ
Protein A/G Purified
RPA-T4
Flow Cytometry
FITC
RUO
NP_001181945
CD4
Primary
Store at 2-4°C.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.