missing translation for 'onlineSavingsMsg'
Learn More

CD4 Mouse, PerCP-Cy5.5, Clone: RPA-T4, enQuireBio™

Mouse Monoclonal Antibody

Brand:  EnquireBio QAB12-PCP55-100Tests

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15931518

  • 228.00 € / 100 tests
Estimated Shipment: 28-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

CD4
Monoclonal
5μL/Test
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2
CD4
Protein A/G Purified
RUO
920
IgG1 κ
Flow Cytometry
RPA-T4
PerCP-Cyanine5.5
NP_001181945
Mouse
100 Tests
Primary
Store at 2-4°C.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions