Learn More
CD4 Rat, PerCP-Cy5.5, Clone: GK1.5, enQuireBio™
Rat Monoclonal Antibody
Brand:  EnquireBio QAB8-PCP55-100ug
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 15932518
  • 305.00 € / 100µg
Estimated Shipment: 27-01-2023
to see stock.
Specifications
CD4
Monoclonal
0.5mg/mL
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, pH7.2
Cd4
Protein A/G Purified
RUO
12504
IgG2b κ
Flow Cytometry
GK1.5
PerCP-Cyanine5.5
NP_038516
Rat
100 μg
Primary
Store at 2-4°C.