missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD4 Rat, V450, Clone: GK1.5, enQuireBio™
Rat Monoclonal Antibody
Brand: EnquireBio QAB8-V450-100ug
This item is not returnable.
View return policy
Specifications
| CD4 | |
| Monoclonal | |
| 0.5mg/mL | |
| 10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, pH7.2 | |
| Cd4 | |
| Protein A/G Purified | |
| RUO | |
| 12504 | |
| IgG2b κ |
| Flow Cytometry | |
| GK1.5 | |
| Violet Fluor 450 (V450) | |
| NP_038516 | |
| Rat | |
| 100 μg | |
| Primary | |
| Store at 2-4°C. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction