missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CD40 Monoclonal Antibody (5C3), NovaFluor™ Yellow 755, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
178.00 € - 392.00 €
Specifications
| Antigen | CD40 |
|---|---|
| Clone | 5C3 |
| Concentration | 0.6 μg/Test |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30055621
|
Invitrogen™
H027T03Y08-A |
100 Tests |
392.00 €
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30055200
|
Invitrogen™
H027T02Y08-A |
25 Tests |
178.00 €
Pack of 25 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD40 Monoclonal antibody specifically detects CD40 in Human samples. It is validated for Flow CytometrySpecifications
| CD40 | |
| 0.6 μg/Test | |
| Monoclonal | |
| Mouse | |
| Human | |
| P25942 | |
| 958 | |
| Primary | |
| 4°C, store in dark, DO NOT FREEZE! |
| 5C3 | |
| Flow Cytometry | |
| NovaFluor Yellow 755 | |
| RUO | |
| PBS with 0.09% sodium azide; pH 7.2 | |
| AI326936; B cell surface antigen CD40; B cell-associated molecule; B-cell surface antigen CD40; Bp50; CD antigen CD40; Cd40; CD40 antigen; CD40 antigen (TNF receptor superfamily member 5); CD40 antigen, TNF receptor superfamily member 5; CD40 molecule; CD40 molecule, TNF receptor superfamily member 5; CD40L receptor; CDw40; GP39; HIGM1; I79_006806; IGM; IMD3; Immunoglobulin M; ImmunoglobulinM; membrane protein CD40; MGC9013; p50; receptor for ligand CD154; sCD40; soluble CD40; T-BAM; T-cell differentiation antigen; Tnfrsf5; TRAP; tumor necrosis factor receptor superfamily member 5; tumor necrosis factor receptor superfamily, member 5 | |
| IgG1 κ | |
| Affinity chromatography | |
| CD40 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title