missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD40/TNFRSF5 Antibody (G28.5) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-52697-0.025MG
This item is not returnable.
View return policy
Description
CD40/TNFRSF5 Monoclonal specifically detects CD40/TNFRSF5 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Functional, CyTOF-ready.
Specifications
| CD40/TNFRSF5 | |
| Monoclonal | |
| Unconjugated | |
| PBS with 0.02% Proclin 300. with 0.02% Proclin 300 | |
| CD40 | |
| Human erythrocyte-rosette-negative tonsillar lymphocytes. | |
| 0.025 mg | |
| Adaptive Immunity, Apoptosis, Asthma, B Cell Development and Differentiation Markers, Cancer, Hematopoietic Stem Cell Markers, Immunology, Signal Transduction, Stem Cell Markers | |
| 958 | |
| Human | |
| Purified |
| Flow Cytometry, Immunohistochemistry, Immunoprecipitation, CyTOF | |
| G28.5 | |
| Flow Cytometry 1:10 - 1:1000, Immunohistochemistry 1:10 - 1:500, Immunoprecipitation 1:10 - 1:500, CyTOF-ready | |
| B cell surface antigen CD40, B-cell surface antigen CD40, Bp50B cell-associated molecule, CD40 antigen, CD40 molecule, TNF receptor superfamily member 5, CD40 type II isoform, CD40L receptor, CDw40, MGC9013, nerve growth factor receptor-related B-lymphocyte activation molecule, p50, TNFRSF5CD40 antigen (TNF receptor superfamily member 5), tumor necrosis factor receptor superfamily member 5, tumor necrosis factor receptor superfamily, member 5 | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| 48 kD Type I glycoprotein, a member of the TNFR superfamily primarily expressed on normal or malignant B cells or B cell lines, macrophages, follicular dendritic cells, endothelial cells, fibroblasts, and at low levels on plasma cells. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction