missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CD41a Monoclonal Antibody (eBioMWReg30 (MWReg30)), NovaFluor™ Blue 555, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
178.00 € - 344.00 €
Specifications
| Antigen | CD41a |
|---|---|
| Clone | eBioMWReg30 (MWReg30) |
| Concentration | 0.1 mg/mL |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30055004
|
Invitrogen™
M044T03B03-A |
100 μg |
344.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30055564
|
Invitrogen™
M044T02B03-A |
25 μg |
178.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD41a Monoclonal antibody specifically detects CD41a in Mouse samples. It is validated for Flow CytometrySpecifications
| CD41a | |
| 0.1 mg/mL | |
| Monoclonal | |
| Rat | |
| Mouse | |
| Q9QUM0 | |
| 16399 | |
| Primary | |
| 4°C, store in dark, DO NOT FREEZE! |
| eBioMWReg30 (MWReg30) | |
| Flow Cytometry | |
| NovaFluor Blue 555 | |
| RUO | |
| PBS with 0.09% sodium azide; pH 7.2 | |
| AI172977; alpha IIb; alphaIIb; alphaIIb protein; BDPLT16; BDPLT2; CD41; CD41B; form 1; form 2; GP IIb; GP2B; GPalpha IIb; GPIIb; GT; GTA; HPA3; integrin alpha 2b; integrin alpha 2b (Cd41b); integrin alpha-IIb; Integrin alpha-IIb heavy chain; Integrin alpha-IIb light chain; Integrin alpha-IIb light chain, form 1; Integrin alpha-IIb light chain, form 2; integrin subunit alpha 2b; integrin subunit alpha IIb; integrin, alpha 2B; integrin, alpha 2b (platelet glycoprotein IIb of IIb/IIIa complex, antigen CD41); ITGA2B; ITGAB; platelet fibrinogen receptor, alpha subunit; platelet glycoprotein IIb; platelet glycoprotein IIb of IIb/II Ia complex; platelet glycoprotein IIb of IIb/IIIa complex; platelet membrane glycoprotein IIb; platelet-specific antigen BAK; PPP1R93; protein phosphatase 1, regulatory subunit 93 | |
| IgG1 κ | |
| Affinity chromatography | |
| Itga2b |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title