missing translation for 'onlineSavingsMsg'
Learn More

CD42b Mouse anti-Human, FITC, Clone: HIP1, BD

Mouse Monoclonal Antibody

81.72 € - 217.00 €

Specifications

Antigen CD42b
Clone HIP1
Applications Flow Cytometry
Classification Monoclonal
Conjugate FITC
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15801409
View Documents
BD Biosciences
561855
25 Tests
ONLINE EXCLUSIVE PRICE
86.70 €
81.72 € / Each
Save 4.98 €
5% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
15838198
View Documents
BD Biosciences
555472
100 Tests
217.00 €
Each
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The HIP1 monoclonal antibody specifically binds to CD42b. CD42b is also known as the Platelet glycoprotein Ib alpha chain that is encoded by the GP1BA gene. CD42b is disulfide bonded to CD42c to form a 170 kDa heterodimer, GPIb. GPIb forms a noncovalent complex with CD42a and CD42d (CD42 complex) that is expressed on platelets and megakaryocytes. The CD42 complex serves as the von Willebrand Factor(vWF) surface receptor involved in the adhesion of platelets to the subendothelium of damaged vascular walls. HIP1 inhibits the ristocetin-dependent binding ofvWF to platelets and partially inhibits collagen-induced aggregation.

Specifications

Specifications

CD42b
Flow Cytometry
FITC
RUO
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
HIP1
Monoclonal
Mouse
Human
IgG1 κ
Affinity Purified
GP1BA; GPIbalpha; GP-Ib alpha; GPIb-alpha; CD42b-alpha; Glycocalicin; BSS
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.