missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CD43 Monoclonal Antibody (eBio84-3C1 (84-3C1)), NovaFluor™ Blue 585, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
178.00 € - 392.00 €
Specifications
| Antigen | CD43 |
|---|---|
| Clone | eBio84-3C1 (84-3C1) |
| Concentration | 0.4 μg/Test |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30055476
|
Invitrogen™
H069T03B04-A |
100 Tests |
392.00 €
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30054948
|
Invitrogen™
H069T02B04-A |
25 Tests |
178.00 €
Pack of 25 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD43 Monoclonal antibody specifically detects CD43 in Human samples. It is validated for Flow CytometrySpecifications
| CD43 | |
| 0.4 μg/Test | |
| Monoclonal | |
| Mouse | |
| Human | |
| P16150 | |
| 6693 | |
| Primary | |
| 4°C, store in dark, DO NOT FREEZE! |
| eBio84-3C1 (84-3C1) | |
| Flow Cytometry | |
| NovaFluor Blue 585 | |
| RUO | |
| PBS with 0.09% sodium azide; pH 7.2 | |
| 3E8 antigen; A630014B01Rik; B-cell differentiation antigen LP-3; CD43; CD43 cytoplasmic tail; CD43-ct; Galactoglycoprotein; GALGP; gpL115; Leukocyte sialoglycoprotein; leukosialin; leukosianin; LOC101120462; LOW QUALITY PROTEIN: leukosialin; Lsn; Lsn1; Ly48; Ly-48; Lymphocyte antigen 48; sialophorin; sialophorin (gpL115, leukosialin, CD43); sialophorin gpL115; SPN; W3/13 antigen | |
| IgG1 κ | |
| Affinity chromatography | |
| SPN |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title