missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PRDX3 Recombinant Rabbit Monoclonal Antibody (24GB935)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA557560
This item is not returnable.
View return policy
Description
PRDX3 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
Peroxiredoxin (Prx) is a growing peroxidase family, whose mammalian members have been known to connect with cell proliferation, differentiation, and apoptosis. Many isoforms (about 50 proteins), collected in accordance to the amino acid sequence homology, containing active site cysteine residue, and the thiol-specific antioxidant activity, distribute throughout all the kingdoms. Among them, mammalian Prx consists of 6 different members grouped into typical 2-Cys, atypical 2-Cys Prx, and 1-Cys Prx. Except Prx VI belonging to 1-Cys Prx subgroup, the other five 2-Cys Prx isotypes have the thioredoxin-dependent peroxidase (TPx) activity utilizing thioredoxin, thioredoxin reductase, and NADPH as a reducing system. Mammalian Prxs are 20-30 kilodalton in molecular size and vary in subcellular localization: Prx I, II, and VI in cytosol, Prx III in mitochondria, Prx IV in ER and secretion, Prx V showing complicated distribution including peroxisome, mitochondria and cytosol.
Specifications
| PRDX3 | |
| Recombinant Monoclonal | |
| Unconjugated | |
| PRDX3 | |
| antioxidant protein; antioxidant protein 1; anti-oxidant protein 1; Aop1; AOP-1; AW822249; D0Tohi1; Ef2l; HBC189; Mer5; mitochondrial thioredoxin dependent peroxide reductase; mitochondrial Trx dependent peroxide reductase; perioredoxin-3; peroxiredoxin 3; peroxiredoxin III; Peroxiredoxin-3; Prdx3; PRO1748; Protein MER5; Protein MER5 homolog; Prx; Prx 3; PRx III; Prx3; PRX-3; Prx-III; SP22; SP-22; TDXM; thioredoxin-dependent peroxide reductase, mitochondrial | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 10935 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 24GB935 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P30048 | |
| PRDX3 | |
| A synthesized peptide derived from human Peroxiredoxin 3 (aa 200-256). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction