missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CD44 Monoclonal Antibody (IM7), NovaFluor™ Red 725, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
178.00 € - 392.00 €
Specifications
| Antigen | CD44 |
|---|---|
| Clone | IM7 |
| Concentration | 0.1 mg/mL |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30055574
|
Invitrogen™
M010T03R05-A |
100 μg |
392.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30055818
|
Invitrogen™
M010T02R05-A |
25 μg |
178.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD44 Monoclonal antibody specifically detects CD44 in Human, Mouse samples. It is validated for Flow CytometrySpecifications
| CD44 | |
| 0.1 mg/mL | |
| Monoclonal | |
| Rat | |
| Human, Mouse | |
| P15379, P16070 | |
| 12505, 960 | |
| Primary | |
| 4°C, store in dark, DO NOT FREEZE! |
| IM7 | |
| Flow Cytometry | |
| NovaFluor Red 725 | |
| RUO | |
| PBS with 0.09% sodium azide; pH 7.2 | |
| AU023126; AW121933; AW146109; Bovine CD44; CD44; CD44 antigen; CD44 antigen-like protein; CD44 molecule; CD44 molecule (Indian blood group); CD44 protein; CD44 variant; CD44A; CD44-like protein; CDW44; cell surface glycoprotein CD44; Cell surface glycoprotein CD44 (hyaluronate binding protein); cell-surface glycoprotein; chondroitin sulfate proteoglycan 8; CSPG8; ECMRIII; ECMR-III; Epican; extracellular matrix receptor III; extracellular matrix receptor-III; GP90 lymphocyte homing/adhesion receptor; HCELL; hematopoietic cell E- and L-selectin ligand; heparan sulfate proteoglycan; HERMES; Hermes antigen; homing function and Indian blood group system; HUTCH-I; hyaluronan receptor; Hyaluronate receptor; IN; LHR; Ly-24; lymphocyte antigen 24; lymphocyte surface antigen precursor CD44; MC56; MDU2; MDU3; METAA; MIC4; PGP I; Pgp1; PGP-1; PGPI; PGP-I; Phagocytic glycoprotein 1; phagocytic glycoprotein I; putative Bos taurus CD44; RHAMM; sCD 44; sCD44; soluble CD 44; soluble CD44 | |
| IgG2b κ | |
| Affinity chromatography | |
| CD44 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title