missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD45 Antibody (5C16) - Azide and BSA Free, Novus Biologicals™
Rat Monoclonal Antibody has been used in 7 publications
213.00 € - 531.00 €
Specifications
| Antigen | CD45 |
|---|---|
| Clone | 5C16 |
| Applications | Immunoprecipitation, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunocytochemistry, Immunofluorescence |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18291162
|
Novus Biologicals
NB110-93609-0.025MG |
0.025 mg |
213.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18778133
|
Novus Biologicals
NB110-93609 |
0.1 mg | |||||||
Description
CD45 Monoclonal specifically detects CD45 in Mouse samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.Specifications
| CD45 | |
| Immunoprecipitation, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| Rat | |
| Adaptive Immunity, Cell Biology, Cellular Markers, Hematopoietic Stem Cell Markers, Immunology, Innate Immunity, Mesenchymal Stem Cell Markers, Microglia Markers, Neuronal Cell Markers, Neuroscience, Signal Transduction, Stem Cell Markers | |
| P06800 | |
| 5788 | |
| Cell lysates of mouse leukocytes | |
| Primary | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
| 5C16 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Mouse | |
| B220, CD45 antigen, CD45R, EC 3.1.3.48, L-CA, LY5, protein tyrosine phosphatase, receptor type, C, receptor-type tyrosine-protein phosphatase C, T200 glycoprotein, T200 leukocyte common antigen, T200receptor type, c polypeptide | |
| PTPRC | |
| IgG2 | |
| Reconstitute with 0.5 ml sterilized PBS to a final concentration of 0.2 mg/ml. | |
| This was selected for its ability to detect mouse CD45 positive leukocytes in IHC. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title