missing translation for 'onlineSavingsMsg'
Learn More

CD45 / LCA Mouse, Clone: PTPRC/1461, enQuireBio™

Product Code. 15990748
Change view
Click to view available options
Quantity:
100 μg
20 μg
Unit Size:
100µg
20µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15990748 100 μg 100µg
15980748 20 μg 20µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15990748 Supplier EnquireBio Supplier No. 5788MSM21P1

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Specifications

Antigen CD45 / LCA
Applications Immunohistochemistry
Cellular Localization Cell surface and cytoplasmic
Classification Monoclonal
Clone PTPRC/1461
Concentration 200μg/mL
Conjugate Unconjugated
Formulation 10mM PBS with 0.05% BSA and 0.05% Azide
Gene Accession No. NP_002829
Gene Symbols PTPRC
Host Species Mouse
Immunogen Human B-lymphocytes
Molecular Weight of Antigen 180-220kDa
Positive Control Ramos, U-698, or GA-10 cells. Tonsil
Purification Method Protein A/G Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5788
Content And Storage Short term: Store at 2-4°C. Long term, aliquot and store at -20°C.
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.