missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD45 / LCA Mouse, Clone: PTPRC/1461, enQuireBio™
Specifications
Specifications
| Antigen | CD45 / LCA |
| Applications | Immunohistochemistry |
| Cellular Localization | Cell surface and cytoplasmic |
| Classification | Monoclonal |
| Clone | PTPRC/1461 |
| Concentration | 200μg/mL |
| Conjugate | Unconjugated |
| Formulation | 10mM PBS with 0.05% BSA and 0.05% Azide |
| Gene Accession No. | NP_002829 |
| Gene Symbols | PTPRC |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?