missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD45 Mouse anti-Human, APC-R700, Clone: HI30, BD Horizon™
Mouse Monoclonal Antibody
352.67 € - 440.84 €
Specifications
| Antigen | CD45 |
|---|---|
| Clone | HI30 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Conjugate | APC-R700 |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15854709
|
BD Biosciences
566042 |
25 Tests |
368.00 € 352.67 € / Each Save 15.33 € 4% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15844709
|
BD Biosciences
566041 |
100 Tests |
458.00 € 440.84 € / Each Save 17.16 € 3% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD45 Monoclonal antibody specifically detects CD45 in Human samples. It is validated for Flow cytometry.
CD45 Monoclonal antibody specifically detects CD45 in Human samples. It is validated for Flow cytometry.Specifications
| CD45 | |
| Flow Cytometry | |
| APC-R700 | |
| Mouse | |
| Cell Biology | |
| Aqueous buffered solution containing BSA, protein stabilizer, glycerol and ≤0.09% sodium azide. | |
| Human Peripheral Blood Leucocytes | |
| Primary | |
| Vol. Per test: 5μL |
| HI30 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| PTPRC; LCA; L-CA; Leukocyte Common Antigen; T200; GP180; LY5 | |
| IgG1 κ | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon APC-R700 under optimum conditions and unconjugated antibody and free BD Horizon APC-R700 were removed. |
For Research Use Only.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title