missing translation for 'onlineSavingsMsg'
Learn More

CD45 Mouse, APC, Clone: HI30, enQuireBio™

Product Code. 15902078
Change view
Click to view available options
Quantity:
100 Tests
25 Tests
500 Tests
Unit Size:
100 tests
25 tests
500 tests
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15902078 100 Tests 100 tests
15912078 25 Tests 25 tests
15922078 500 Tests 500 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15902078 Supplier EnquireBio Supplier No. QAB46APC100Tests

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Specifications

Antigen CD45
Applications Flow Cytometry
Classification Monoclonal
Clone HI30
Concentration 5μL/Test
Conjugate APC
Formulation 10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2
Gene Accession No. NP_002829
Gene Symbols PTPRC
Host Species Mouse
Purification Method Protein A/G Purified
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5788
Content And Storage Store at 2-4°C.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.