missing translation for 'onlineSavingsMsg'
Learn More

CD45 Mouse, APC-Cy7, Clone: HI30, enQuireBio™

Mouse Monoclonal Antibody

Brand:  EnquireBio QAB46-APC7-100Tests

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15932078

  • 295.00 € / 100 tests
Estimated Shipment: 13-11-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

CD45
Monoclonal
5μL/Test
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2
PTPRC
Protein A/G Purified
RUO
5788
IgG1 κ
Flow Cytometry
HI30
APC-Cyanine7
NP_002829
Mouse
100 Tests
Primary
Store at 2-4°C.
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers