missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD45 Mouse, PE-Cy7, Clone: HI30, enQuireBio™
Mouse Monoclonal Antibody
196.35 € - 367.50 €
Specifications
| Antigen | CD45 |
|---|---|
| Clone | HI30 |
| Concentration | 5μL/Test |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15932088
|
EnquireBio
QAB46-PE7-25Tests |
25 Tests |
196.35 €
25 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15922088
|
EnquireBio
QAB46-PE7-100Tests |
100 Tests |
367.50 €
100 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
| CD45 | |
| 5μL/Test | |
| Monoclonal | |
| Mouse | |
| 10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2 | |
| 5788 | |
| IgG1 κ | |
| Protein A/G Purified |
| HI30 | |
| Flow Cytometry | |
| PE-Cyanine7 | |
| RUO | |
| NP_002829 | |
| PTPRC | |
| Primary | |
| Store at 2-4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title