missing translation for 'onlineSavingsMsg'
Learn More

CD45 Mouse, PerCP-Cy5.5, Clone: 2D1, enQuireBio™

Mouse Monoclonal Antibody

327.60 € - 3202.50 €

Specifications

Antigen CD45
Clone 2D1
Concentration 5μL/Test
Applications Flow Cytometry
Classification Monoclonal
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15902658
View Documents
EnquireBio
QAB97-PCP55-25Tests
25 Tests
327.60 €
25 tests
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
15992648
View Documents
EnquireBio
QAB97-PCP55-100Tests
100 Tests
738.15 €
100 tests
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
15912658
View Documents
EnquireBio
QAB97-PCP55-500Tests
500 Tests
3202.50 €
500 tests
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Specifications

CD45
5μL/Test
Monoclonal
Mouse
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2
5788
IgG1 κ
Protein A/G Purified
2D1
Flow Cytometry
PerCP-Cyanine5.5
RUO
NP_002829
PTPRC
Primary
Store at 2-4°C.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.