missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD45 Rabbit anti-Human, Unconjugated, Clone: 6O19, Proteintech
Rabbit Recombinant Antibody
149.00 € - 439.00 €
Specifications
| Antigen | CD45 |
|---|---|
| Clone | 6O19 |
| Applications | Western Blot, Immunohistochemistry, Flow Cytometry |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17551496
|
Proteintech
80297-1-RR-20UL |
20 μL |
149.00 €
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17541496
|
Proteintech
80297-1-RR-100UL |
100 μL |
439.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The CD45 antibody from Proteintech is a rabbit recombinant antibody to a recombinant protein of human CD45. This antibody recognizes human, rat antigen. The CD45 antibody has been validated for the following applications: FC, IHC, WB, ELISA analysis.
Specifications
| CD45 | |
| Western Blot, Immunohistochemistry, Flow Cytometry | |
| Rabbit | |
| 5788 | |
| Primary |
| 6O19 | |
| Unconjugated | |
| Human, Rat | |
| IgG |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title