missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD45 Rat anti-Mouse, BB700, Clone: 30-F11, BD Horizon™
Description
CD45 Monoclonal antibody specifically detects CD45 in Mouse samples. It is validated for Flow Cytometry.
Specifications
Specifications
| Antigen | CD45 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | 30-F11 |
| Concentration | 0.2 mg/mL |
| Conjugate | Brilliant Blue 700 (BB700) |
| Host Species | Rat |
| Quantity | 25 μg |
| Regulatory Status | RUO |
| Primary or Secondary | Primary |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?