missing translation for 'onlineSavingsMsg'
Learn More

CD45R/B220 Rat anti-Mouse, FITC, Clone: RA3-6B2, BD

Rat Monoclonal Antibody

Brand:  BD Biosciences 553088

 View more versions of this product

Product Code. 15876988

  • ONLINE EXCLUSIVE PRICE
    473.00 €
    453.68 € / Each
    Save 19.32 €
    4% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

The RA3-6B2 antibody reacts with an epitope on the extracellular domain of CD45 glycoprotein which is dependent upon the expression of exon A and specific carbohydrate residues. It is expressed on B lyphocytes at all stages from pro-B through mature and activated B cell, but it is decreased on plasma cells and a subset of memory B cells. Levels of expression of CD45R/B220 on the B-cell lineage appear to be developmentally regulated. It is also found on the abnormal T cells involved in the lymphadenopathy of lpr/lpr and gld/gld mutant mice, on lytically active subsets of lymphokine-activated killer cells (NK cells and non-MHC-restricted CTL), on apoptotic T lymphocytes of mice injected with bacterial superantigen, on a population of NK-cell precursors in the bone marrow, and on B-lymphocyte, T-lymphocyte, and macrophage progenitors in fetal liver. The CD45R/B220 antigen is not on hematopoietic stem cells, naive T lymphocytes, or MHC-restricted CTL. CD45 is a member of the Protein Tyrosine Phosphatase (PTP) family: Its intracellular (COOH-terminal) region contains two PTP catalytic domains, and the extracellular region is highly variable due to alternative splicing of exons 4, 5, and 6 (designated A, B, and C, respectively), plus differing levels of glycosylation. The CD45 isoforms detected in the mouse are cell type-, maturation, and activation state-specific. The CD45 isoforms play complex roles in T-cell and B-cell antigen receptor signal transduction. CD45R/B220 is commonly used as a pan B-cell marker; however, CD19 expression, detected by mAb 1D3, is reported to be more restricted to the B-cell lineage. mAb RA3-6B2 has been reported to enhance isotype switching during in vitro B-cell responses and to inhibit in vivo B-cell responses. Cross-reaction of RA3-6B2 mAb with activated human T lymphocytes has been observed. This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.

Host Species: Rat
Clone: RA3-6B2
Isotype: IgG2a κ
Species Reactivity: Mouse
Immunogen: Mouse Pre-B Tumor

Flow Cytometry, Immunofluorescence

TRUSTED_SUSTAINABILITY
Specifications

Specifications

CD45R
Monoclonal
0.5mg/mL
Aqueous buffered solution containing ≤0.09% sodium azide.
Mouse Pre-B Tumor
0.5 mg
Immunology
Human, Mouse
IgG2a κ
Flow Cytometry
RA3-6B2
FITC
Rat
Affinity Purified
RUO
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only.