missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD45RA Antibody (T200/8144R), DyLight 350, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-20794UV
619.65 EUR valid until 2025-12-16
BEST PRICE on Promo! Use promo code "24090" to get your promotional price.
This item is not returnable.
View return policy
Description
CD45RA Monoclonal antibody specifically detects CD45RA in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| CD45RA | |
| Monoclonal | |
| DyLight 350 | |
| B220, CD45 antigen, CD45R, EC 3.1.3.48, L-CA, LY5, protein tyrosine phosphatase, receptor type, C, receptor-type tyrosine-protein phosphatase C, T200 glycoprotein, T200 leukocyte common antigen, T200receptor type, c polypeptide | |
| Recombinant full-length human CD45RA protein | |
| 0.1 mL | |
| Adaptive Immunity, Cell Biology, Cellular Markers, Cellular Signaling, Cytokine Research, Glia Markers, Hematopoietic Stem Cell Markers, Immunology, MAP Kinase Signaling, Mast Cell Markers, Mesenchymal Stem Cell Markers, Microglia Markers, Modulation of DNA Pools, Myeloid Cell Markers, Myeloid derived Suppressor Cell, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Signal Transduction, Stem Cell Markers | |
| 5788 | |
| Store at 4°C in the dark. | |
| IgG κ |
| Immunohistochemistry (Paraffin) | |
| T200/8144R | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction