missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CD45RA Monoclonal Antibody (HI100), NovaFluor™ Red 710, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
178.00 € - 392.00 €
Specifications
| Antigen | CD45RA |
|---|---|
| Clone | HI100 |
| Concentration | 0.8 μg/Test |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30055449
|
Invitrogen™
H014T03R04-A |
100 Tests |
392.00 €
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30056043
|
Invitrogen™
H014T02R04-A |
25 Tests |
178.00 €
Pack of 25 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD45RA Monoclonal antibody specifically detects CD45RA in Human samples. It is validated for Flow CytometrySpecifications
| CD45RA | |
| 0.8 μg/Test | |
| Monoclonal | |
| Mouse | |
| Human | |
| P08575 | |
| 5788 | |
| Primary | |
| 4°C, store in dark, DO NOT FREEZE! |
| HI100 | |
| Flow Cytometry | |
| NovaFluor Red 710 | |
| RUO | |
| PBS with 0.09% sodium azide; pH 7.2 | |
| B220; CD45; CD45 antigen; CD45 antigen isoform 1 precursor; CD45 antigen isoform 2 precursor; CD45 antigen isoform 3 precursor; CD45 antigen isoform 4 precursor; CD45 antigen isoform 5 precursor; CD45 antigen isoform 6 precursor; CD45R; GP180; LCA; L-CA; leucocyte common antigen; Leukocyte common antigen; leukocyte common antigen A; leukocyte common antigen B; LOW QUALITY PROTEIN: receptor-type tyrosine-protein phosphatase C; LY5; Ly-5; Lyt-4; protein tyrosine phosphatase receptor type C; protein tyrosine phosphatase, receptor type C; protein tyrosine phosphatase, receptor type, C; protein tyrosine phosphatase, receptor type, c polypeptide; Protein tyrosine phosphatase, receptor-type, c polypeptide; protein tyrosine phosphatase; alternatively spliced; PTPRC; Receptor-type tyrosine-protein phosphatase C; RT7; T100; T200; T200 glycoprotein; T200 leukocyte common antigen | |
| IgG2b κ | |
| Affinity chromatography | |
| PTPRC |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title