Learn More
CD45RA Mouse anti-Rat, PE, Clone: OX-33, BD
Mouse Monoclonal Antibody
398.40 € - 567.10 €
Specifications
| Antigen | CD45RA |
|---|---|
| Clone | OX-33 |
| Concentration | 0.2mg/mL |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15806648
|
BD Biosciences
551402 |
0.1 mg |
415.00 € 398.40 € / Each Save 16.60 € 4% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
10463085
|
BD Biosciences
554884 |
0.2 mg |
595.00 € 567.10 € / Each Save 27.90 € 4% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The OX-33 antibody specifically recognizes a high-molecular-weight form of CD45 found only on B lymphocytes. CD45 is a member of the Protein Tyrosine Phosphatase (PTP) family: Its intracellular (COOH-terminal) region contains two PTP catalytic domains, and the extracellular region is highly variable due to alternative splicing of exons 4, 5, and 6 (designated A, B, and C, respectively), plus differing levels of glycosylation. The CD45 isoforms detected in the rat are cell type-, maturation-, and activation state-specific. The CD45 isoforms play complex roles in T-cell and B-cell antigen receptor signal transduction.
This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
Specifications
| CD45RA | |
| 0.2mg/mL | |
| Monoclonal | |
| Mouse | |
| Immunology | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| IgG1 κ | |
| Affinity Purified |
| OX-33 | |
| Flow Cytometry | |
| PE | |
| RUO | |
| Rat | |
| Leukocyte common antigen purified from rat splenocytes | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.