missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD45RO Antibody (190-2F2.5), Biotin, Novus Biologicals™
Mouse Monoclonal Antibody
553.00 € - 598.00 €
Specifications
| Antigen | CD45RO |
|---|---|
| Clone | 190-2F2.5 |
| Applications | Flow Cytometry, Immunofluorescence, Immunofluorescence, CyTOF |
| Classification | Monoclonal |
| Conjugate | Biotin |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18627846
|
Novus Biologicals
NBP2-47956B-100ul |
100 μg |
598.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18604707
|
Novus Biologicals
NBP2-47956B |
0.1 mL |
553.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD45RO Monoclonal antibody specifically detects CD45RO in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/ Immunofluorescence, Immunofluorescence, CyTOFSpecifications
| CD45RO | |
| Flow Cytometry, Immunofluorescence, Immunofluorescence, CyTOF | |
| Biotin | |
| Mouse | |
| Adaptive Immunity, Cell Biology, Cellular Markers, Cytokine Research, Hematopoietic Stem Cell Markers, Immunology, Mesenchymal Stem Cell Markers, Microglia Markers, Neuronal Cell Markers, Neuroscience, Signal Transduction, Stem Cell Markers | |
| PBS | |
| 5788 | |
| IgG2a κ | |
| Protein A or G purified |
| 190-2F2.5 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| B220, CD45 antigen, CD45R, EC 3.1.3.48, L-CA, LY5, protein tyrosine phosphatase, receptor type, C, receptor-type tyrosine-protein phosphatase C, T200 glycoprotein, T200 leukocyte common antigen, T200receptor type, c polypeptide | |
| Lentillectin Purified glyco proteins obtained from HuPBL and from a CLL (B cell). | |
| Primary | |
| Store at 4°C in the dark. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title