Learn More
CD45RO Antibody (ILA116A), Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-61035
Product Code. 18258654
  • 332.00 € / 0.10mg
Description
CD45RO Monoclonal specifically detects CD45RO in Bovine, Goat, Water Buffalo, Sheep samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CD45RO
Monoclonal
1 mg/mL
Flow Cytometry 1:10 - 1:1000
B220, CD45 antigen, CD45R, EC 3.1.3.48, L-CA, LY5, protein tyrosine phosphatase, receptor type, C, receptor-type tyrosine-protein phosphatase C, T200 glycoprotein, T200 leukocyte common antigen, T200receptor type, c polypeptide
Mouse
Unpurified
Adaptive Immunity, Cell Biology, Cellular Markers, Cellular Signaling, Cytokine Research, Glia Markers, Hematopoietic Stem Cell Markers, Immunology, Immunophenotyping, MAP Kinase Signaling, Mast Cell Markers, Mesenchymal Stem Cell Markers, Microglia Markers, Modulation of DNA Pools, Myeloid Cell Markers, Myeloid derived Suppressor Cell, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Signal Transduction, Stem Cell Markers
5788
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Flow Cytometry, Immunohistochemistry, Functional Assay
ILA116A
Unconjugated
Ascites with 0.09% Sodium Azide
PTPRC
Bovine mononuclear leukocytes
0.1 mg
Primary
Bovine, Goat, Water Buffalo, Sheep, Feline, Hamster
IgG3