missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD46 Antibody (MCP/931), Alexa Fluor™ 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-11539AF350
This item is not returnable.
View return policy
Description
CD46 Monoclonal antibody specifically detects CD46 in Human samples. It is validated for Flow Cytometry, Immunofluorescence
Specifications
| CD46 | |
| Monoclonal | |
| Alexa Fluor 350 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, Immunofluorescence | |
| MCP/931 | |
| Flow Cytometry, Immunocytochemistry/Immunofluorescence | |
| AHUS2, CD46 antigen, CD46 molecule, complement regulatory protein, membrane cofactor protein (CD46, trophoblast-lymphocyte cross-reactive antigen), MIC10, TLXcomplement regulatory protein, trophoblast leucocyte common antigen, Trophoblast leukocyte common antigen, trophoblast-lymphocyte cross-reactive antigen | |
| Recombinant human CD46 protein (Uniprot: P15529) | |
| 0.1 mL | |
| Cancer, Cell Biology, Cellular Markers, Immunology | |
| 4179 | |
| Store at 4°C in the dark. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction